Anti-SMAD6 Mouse Monoclonal Antibody [clone: 1G2]

Leverantör: United States Biological
USBIS1014-55H7EA 8175 SEK
USBIS1014-55H7
Anti-SMAD6 Mouse Monoclonal Antibody [clone: 1G2]
Antikroppar
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: SMAD6 (SMAD family member 6)
Clonality: Monoclonal
Clone: 1G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR