Anti-SMAD7 Rabbit Polyclonal Antibody

Leverantör: United States Biological
USBIS1014-55M3EA 6675 SEK
USBIS1014-55M3
Anti-SMAD7 Rabbit Polyclonal Antibody
Antikroppar
Recommended dilution: Western Blot: 1:500-1:1000

Type: Primary
Antigen: SMAD7 (SMAD family member 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR