Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Anti-SMAGP Goat polyclonal antibody unconjugated

Leverantör: US Biological
Recommended dilution: Western Blot: 0.1-0.2μg/ml; ELISA: 0.5-1 μg/ml

Type: Primary
Antigen: SMAGP (small cell adhesion glycoprotein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat




 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
100 µg 100 µG USBIS1014-55V
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol SMAGP
Antigen name Small cell adhesion glycoprotein
Reaktivitet Rat
Conjugation Unconjugated
Clonality Polyclonal
Antikroppstyp Primary
Värd Goat
Isotype IgG
Western blot Yes
ELISA Yes
Aminosyretal 56 to 97
Rening purified by Protein G chromatography
Form lyophilized powder
Storage buffer PBS, 5% trehalose.
Förvaringstemperatur Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal