Anti-Small Glutamine-rich Tetratricopeptide Chicken Polyclonal Antibody
Leverantör: United States Biological
USBIS1014-58PEA 7325 SEK
USBIS1014-58P
Anti-Small Glutamine-rich Tetratricopeptide Chicken Polyclonal Antibody
Antikroppar
Recommended dilution: Western Blot: 1:500; Immunochemistry: 1:200
Type: Primary
Antigen: Small Glutamine-rich Tetratricopeptide
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human