Anti-Small Inducible Cytokine Subfamily A Chicken Polyclonal Antibody

Leverantör: United States Biological
USBIS1014-58Q1EA 5525 SEK
USBIS1014-58Q1
Anti-Small Inducible Cytokine Subfamily A Chicken Polyclonal Antibody
Antikroppar
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: Small Inducible Cytokine Subfamily A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR