Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Anti-Small Inducible Cytokine Subfamily E, Member 1 Chicken polyclonal antibody unconjugated

Leverantör: US Biological
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: Small Inducible Cytokine Subfamily E, Member 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human




 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
50 µg 50 µG USBIS1014-58R1
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol Small Inducible Cytokine Subfamily E, Member 1
Reaktivitet Human
Conjugation Unconjugated
Clonality Polyclonal
Antikroppstyp Primary
Värd Chicken
Isotype IgY
Western blot Yes
ELISA Yes
Aminosyretal 130 to 201
Rening purified by immunoaffinity chromatography
Form liquid
Storage buffer PBS, pH 7,2, 0,02% sodium azide.
Förvaringstemperatur Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal