Vill du logga in igen?
Anti-HLTF Rabbit polyclonal antibody unconjugated
Leverantör: US Biological
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100
Type: Primary
Antigen: HLTF (helicase-like transcription factor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
ORDER
Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
200 µl 200 µl USBIS1014-58YA
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFIKATIONER
Antigen symbol HLTF
Antigen name Helicase-like transcription factor
Reaktivitet Human
Conjugation Unconjugated
Clonality Polyclonal
Antikroppstyp Primary
Värd Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Rening purified by ammonium sulfate precipitation
Form liquid
Storage buffer PBS, 0,70% sodium azide.
Förvaringstemperatur Cool
Shipping temperature Ice
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal