Anti-SMG1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Leverantör: United States Biological
USBIS1014-58Z1DAPCEA 9900 SEK
USBIS1014-58Z1DAPC
Anti-SMG1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikroppar
Type: Primary
Antigen: SMG1 (SMG1 phosphatidylinositol 3-kinase-related kinase)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR