Anti-SMG1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Leverantör: United States Biological
USBIS1014-58Z1F-APEA 9900 SEK
USBIS1014-58Z1F-AP
Anti-SMG1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikroppar
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: SMG1 (SMG1 phosphatidylinositol 3-kinase-related kinase)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human