Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Anti-SMURF1 Mouse monoclonal antibody unconjugated [clone: 1D7]

Leverantör: US Biological
Recommended dilution: Immunofluorescence: 10 μg/ml; Immunochemistry: 5 μg/ml

Type: Primary
Antigen: SMURF1 (SMAD specific E3 ubiquitin protein ligase 1)
Clonality: Monoclonal
Clone: 1D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human




 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
50 µg 50 µG USBIS1014-60B1
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol SMURF1
Antigen name SMAD specific E3 ubiquitin Protein ligase 1
Reaktivitet Human
Conjugation Unconjugated
Clonality Monoclonal
Antikroppstyp Primary
Clone 1D7
Värd Mouse
Isotype IgG2a kappa
Western blot Yes
ELISA Yes
ImmunoPrecipitation Yes
Immunofluorescence Yes
Aminosyretal 165 to 269
Koncentration ~0,1 mg/ml
Rening purified by Protein A chromatography
Form liquid
Storage buffer PBS, pH 7,2.
Förvaringstemperatur Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal