Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Anti-SMARCD2 Mouse monoclonal antibody unconjugated [clone: 2F7]

Leverantör: US Biological
Recommended dilution: Immunofluorescence: 10 μg/ml; Immunochemistry: 5 μg/ml

Type: Primary
Antigen: SMARCD2 (SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2)
Clonality: Monoclonal
Clone: 2F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human




 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
50 µg 50 µG USBIS1014-61K
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol SMARCD2
Antigen name SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2
Reaktivitet Human
Conjugation Unconjugated
Clonality Monoclonal
Antikroppstyp Primary
Clone 2F7
Värd Mouse
Isotype IgG2a kappa
Western blot Yes
ELISA Yes
Immunofluorescence Yes
Aminosyretal 398 to 475
Koncentration ~0,5 mg/ml
Rening purified by Protein A chromatography
Form liquid
Storage buffer PBS, pH 7,2.
Förvaringstemperatur Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal