Anti-SMURF2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Leverantör: United States Biological
USBIS1014-65E-HRPEA 9900 SEK
USBIS1014-65E-HRP
Anti-SMURF2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Type: Primary
Antigen: SMURF2 (SMAD specific E3 ubiquitin protein ligase 2)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse