Anti-SMURF2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Leverantör: United States Biological
USBIS1014-65E-PEEA 9900 SEK
USBIS1014-65E-PE
Anti-SMURF2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: SMURF2 (SMAD specific E3 ubiquitin protein ligase 2)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse