Anti-SNAI1 Rat Monoclonal Antibody [clone: 7H63]
Leverantör: United States Biological
USBIS1014-67HEA 6450 SEK
USBIS1014-67H
Anti-SNAI1 Rat Monoclonal Antibody [clone: 7H63]
Antikroppar
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: SNAI1 (snail family zinc finger 1)
Clonality: Monoclonal
Clone: 7H63
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Human