Anti-SNAI1 Rabbit Polyclonal Antibody
Leverantör: United States Biological
USBIS1014-67K2EA 6825 SEK
USBIS1014-67K2
Anti-SNAI1 Rabbit Polyclonal Antibody
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:10-1:50
Type: Primary
Antigen: SNAI1 (snail family zinc finger 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human