Anti-SNAI1 Rabbit Polyclonal Antibody

Leverantör: United States Biological
USBIS1014-67LEA 6825 SEK
USBIS1014-67L
Anti-SNAI1 Rabbit Polyclonal Antibody
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: SNAI1 (snail family zinc finger 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR