Anti-SNAI Rabbit polyclonal antibody FITC (Fluorescein)
USBIS1014-67L1FITCEA 5975 SEK
USBIS1014-67L1FITC
Anti-SNAI Rabbit polyclonal antibody FITC (Fluorescein)
Type: Primary
Antigen: SNAI
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human