Anti-SNRPD1 Rabbit Polyclonal Antibody
Leverantör: United States Biological
USBIS1014-96AEA 6825 SEK
USBIS1014-96A
Anti-SNRPD1 Rabbit Polyclonal Antibody
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100
Type: Primary
Antigen: SNRPD1 (small nuclear ribonucleoprotein D1 polypeptide 16kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human