Anti-SNRPD1 Mouse Monoclonal Antibody [clone: 8C306]
Leverantör: United States Biological
USBIS1014-96NEA 4325 SEK
USBIS1014-96N
Anti-SNRPD1 Mouse Monoclonal Antibody [clone: 8C306]
Antikroppar
Recommended dilution: Western Blot: 1:1000-1:2000
Type: Primary
Antigen: SNRPD1 (small nuclear ribonucleoprotein D1 polypeptide 16kDa)
Clonality: Monoclonal
Clone: 8C306
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now