Anti-SNX1 Goat Polyclonal Antibody
Leverantör: United States Biological
USBIS1014-97A6EA 6400 SEK
USBIS1014-97A6
Anti-SNX1 Goat Polyclonal Antibody
Antikroppar
Recommended dilution: ELISA: 1: 64000; Western Blot: 1-3?g/ml
Type: Primary
Antigen: SNX1 (sorting nexin 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human