Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Anti-SNX6 Rabbit polyclonal antibody FITC (Fluorescein)

Leverantör: US Biological
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: SNX6 (sorting nexin 6)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human



Logga in för att se avtalspriser och tillgänglighet.
  Kom ihåg mig i den här enheten
 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
200 µl 200 µl USBIS1014-97C-FITC
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol SNX6
Antigen name sorting nexin 6
Reaktivitet Human
Conjugation FITC (Fluorescein Isothiocyanate)
Clonality Polyclonal
Antikroppstyp Primary
Värd Rabbit
Isotype IgG
Western Blot Yes
ELISA Yes
Epitop N-terminal
Rening purified by Protein A chromatography
Form liquid
Storage Buffer PBS, pH 7,2, contains ~0,09% sodium azide
Storage temperature Cool
Shipping Temperature Ice
Tested Applications Fluoresence ELISA
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Benämning Artikel nummer Availability Enhet Ditt pris Price Per Antal