Anti-SNX7A Chicken Polyclonal Antibody

Leverantör: United States Biological
USBIS1014-97GEA 8475 SEK
USBIS1014-97G
Anti-SNX7A Chicken Polyclonal Antibody
Antikroppar
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: SNX7A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR