Anti-SNX12 Goat Polyclonal Antibody

Leverantör: United States Biological
USBIS1014-97L4EA 6400 SEK
USBIS1014-97L4
Anti-SNX12 Goat Polyclonal Antibody
Antikroppar
Recommended dilution: ELISA: 1:32000; Immunochemistry: 3-6?g/ml

Type: Primary
Antigen: SNX12 (sorting nexin 12)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR