Anti-SNX18 Goat Polyclonal Antibody

Leverantör: United States Biological
USBIS1014-97Q2EA 6400 SEK
USBIS1014-97Q2
Anti-SNX18 Goat Polyclonal Antibody
Antikroppar
Recommended dilution: ELISA: 1:32000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: SNX18 (sorting nexin 18)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR