Anti-SNX19 Goat Polyclonal Antibody

Leverantör: United States Biological
USBIS1014-97R1EA 6400 SEK
USBIS1014-97R1
Anti-SNX19 Goat Polyclonal Antibody
Antikroppar
Recommended dilution: ELISA: 1:16000; Western Blot: 0. 3-1 ?g/ml

Type: Primary
Antigen: SNX19 (sorting nexin 19)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR