Anti-STK19 Rabbit Polyclonal Antibody (Biotin)

Leverantör: United States Biological
USBIS4283-30A-BIOTEA 9900 SEK
USBIS4283-30A-BIOT
Anti-STK19 Rabbit Polyclonal Antibody (Biotin)
Antikroppar
Type: Primary
Antigen: STK19 (serine/threonine kinase 19)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR