Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Anti-STK19 Rabbit polyclonal antibody HRP (Horseradish Peroxidase)

Leverantör: US Biological
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: STK19 (serine/threonine kinase 19)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human




 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
200 µl 200 µl USBIS4283-30A-HRP
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol STK19
Antigen name Serine/threonine Kinase 19
Reaktivitet Human
Conjugation HRP (Horseradish Peroxidase)
Clonality Polyclonal
Antikroppstyp Primary
Värd Rabbit
Isotype IgG
ELISA Yes
Epitop N-terminal
Rening purified by Protein G chromatography
Form liquid
Storage buffer PBS, pH 7,2
Förvaringstemperatur Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping