Anti-SRPK3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Leverantör: United States Biological
USBIS4283-34-APCEA 9900 SEK
USBIS4283-34-APC
Anti-SRPK3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: SRPK3 (SRSF protein kinase 3)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR