Flow meter
Leverantör: PerkinElmer
PERC02901624EA 13150 SEK
PERC02901624
Flow meter
Flow meter designed to measure the nitrogen flow.
  • Control and conserve purge gases
  • Includes 1/8 NPT female connection
Suitable for float displacement.