Petri dishes, extra depth, disposable, Lab-Tek™
Leverantör: Thermo Fisher Scientific
NUNC4031EA 5575 SEK
NUNC4031
Petri dishes, extra depth, disposable, Lab-Tek™
Petri dishes for culturing fungi, bacteria and other microorganisms.
  • Compatible with automated systems
  • Deep Petri dishes allow for longer culture periods
  • Promote larger plaques used for efficient library screening
Förpackning: 12 plates per sealed plastic sleeve.