Differential refractometer, Smartline 2300
Leverantör: Knauer
KNAUA5160EA 0 SEK
KNAUA5160
Differential refractometer, Smartline 2300
Refraktometrar Refraktometrar, bordsmodeller
The Smartline 2300 is a differential refractometer suitable for detecting compounds with little or without UV activity such as sugars, lipids or polymers.
  • High sensitivity
  • Integrated flush valve for rinsing the reference cell
  • No pre-tempering necessary
This detector can be controlled with OpenLab® EZChrom, ChromGate® and ClarityChrom® software via RS232 from the front panel (stand-alone operation), as well as through analog input/output allowing it to be integrated into almost any LC system.
Designed for use in analytical HPLC as well as for GPC applications.
Leveransinformation: Supplied with a 15 L flowcell.