Anti-IgG Goat Monoclonal Antibody (HRP (Horseradish Peroxidase))

Leverantör: Abnova
ABNOPAB10752EA 4250 SEK
ABNOPAB10752
Anti-IgG Goat Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antikroppar
Goat anti-mouse IgG recognizes mouse IgG F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Peroxidase.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: F(ab')2
Host:
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR