Anti-IgG Rabbit Monoclonal Antibody

Leverantör: Abnova
ABNOPAB9172EA 4875 SEK
ABNOPAB9172
Anti-IgG Rabbit Monoclonal Antibody
Antikroppar
Rabbit anti-chicken IgG recognizes chicken IgG F(c) fragment. This secondary antibody was purified using antigen affinity chromatography.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Rabbit
Isotype:
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR