Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Goat Anti-Chicken IgG Monoclonal Antibody (Unconjugated)

Leverantör: Abnova
Goat anti-chicken IgG recognizes chicken IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Chicken




 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
1 mg 1 mg ABNOPAB9169
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol IgG
Antigen name Immunoglobulin G
Reaktivitet Chicken
Conjugation Unconjugated
Clonality Monoclonal
Antikroppstyp Secondary
Värd Goat
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Epitop Heavy and Light chain
Mått Liquid
Cross adsorption No
Immunogen Chicken IgG whole molecule
Rening solid phase adsorption
Storage buffer 20 mM KH2PO4, 150 mM NaCl, pH 7,2 (0,01% sodium azide)
Förvaringstemperatur Store at 4 °C. Do not Freeze.
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal