Anti-IgM µ Goat Monoclonal Antibody

Leverantör: Abnova
ABNOPAB9168EA 4875 SEK
ABNOPAB9168
Anti-IgM µ Goat Monoclonal Antibody
Antikroppar
Goat anti-chicken IgG recognizes chicken IgM mu heavy chain. This secondary antibody was purified using antigen affinity chromatography.

Type: Secondary
Antigen: IgM µ
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope: µ-chain
Host: Goat
Isotype:
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR