Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Goat Anti-Mouse IgG Monoclonal Antibody (HRP (Horseradish Peroxidase))

Leverantör: Abnova
Goat anti-mouse IgG recognizes mouse IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Peroxidase.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Mouse



 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
2 mg 2 mg ABNOPAB10746
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol IgG
Antigen name Immunoglobulin G
Reaktivitet Mouse
Conjugation HRP (Horseradish Peroxidase)
Clonality Monoclonal
Antikroppstyp Secondary
Värd Goat
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Epitop Heavy and Light chain
Mått Lyophilized
Cross adsorption No
Immunogen Mouse IgG whole molecule
Rening solid phase adsorption
Storage buffer 10 mM potassium phosphate, 75 mM NaCl, 50% (v/v) glycerol, pH 7,2 (0,01% (w/v) gentamicsulfate (Do NOT add sodium azide!), 10 mg/mL BSA (Immunoglobuland Protease free))
Förvaringstemperatur Store at 4 °C on dry atmosphere. After reconstitution with 1 mL of deionized water, store at – 20 °C or lower. Aliquot to avoid repeated freezing and thawing.
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping