Full face masks, FM2
Leverantör: Scott Safety
SCTS2023639EA 1900 SEK
SCTS2023639 SCTS2023643
Full face masks, FM2
Reuseable rubber or silicone face mask for use with powered air respiratory Spirit. The full facemask FM2 provides the highest levels of comfort and wearer acceptance​.
  • Natural rubber front-port face mask
  • Large visor area and broad face seal
  • Single size full facemask also approved for unpowered use with Pro2000 filters
Certifikat: EN 12942 TM3 and AS/NZS1716-P3
Observera: *One size fits all