Technical calculator, Casio, FX-82ES
Leverantör: Casio
EDSC127298EA 450 SEK
EDSC127298
Technical calculator, Casio, FX-82ES
Technical calculator with six variable memories and two-dimensional statistics.
  • 249 functions including 160 scientific functions
  • Natural textbook display: 31×96 dot matrix
  • Power supply: One AAA battery
Functions: Two-line dot matrix display - 15 and 10+2 digits - Trigonometric functions - Six variable memories - Two-dimensional statistics - Regression calculation - Repeat function - Natural textbook display - Automatic fraction calculation (2 modes) - Function table.
Leveransinformation: Supplied with a hard case.