Human recombinant GMFB
Leverantör: Tonbo Biosciences
21-7108-U010EA 2075 SEK
TNBO21-7108-U010 TNBO21-7108-U500 TNBO21-7108-U100 TNBO21-7108-U002
Human recombinant GMFB
Glia Maturation Factor beta (GMF-β) is a nerve growth factor and member of the actin-binding proteins ADF family. It is mainly localised in the brain, and functions in the growth and differentiation of neurons and glia. It plays a role in stimulating neural regeneration. GMF-beta has also been found to inhibit the proliferation of a variety of tumor cells in culture.