Vill du logga in igen?
Detergent solutions, Surfact-Amps™
Leverantör: Thermo Scientific
Pierce
Surfact-Amps™ detergents are ready-to-use, diluted solutions of purified detergent that can be used in routine and high-demand protein
research methods and molecular biology techniques. These detergents are useful as a component of cell lysis buffers or other solutions intended to extract and solubilise proteins, improve chromatography, prepare yeast spheroplasts, and reduce background in ELISA or Western blot methods.
  • Clear and pure 10% detergent solutions with <1.0 μeq/ml peroxides and carbonyl content
  • Ampoules are packed under nitrogen for maximum shelf life and are pre-scored for easy opening
  • Easy to pipette and accurately dispense
  • Packaging eliminates the accumulation of peroxides and degradation products
Detergents are a class of molecules whose unique properties enable manipulation (disruption or formation) of hydrophobichydrophilic interactions among molecules in biological samples. In biological research, detergents are used to lyse cells (release soluble proteins), solubilise membrane proteins and lipids, control protein crystallisation and prevent non specific binding in affinity purification and immunoassay
procedures. They are often used as an additive in protein gel electrophoresis.
ORDER
Expandera alla / Komprimera alla
Benämning Förp. VWR art.nr Antal
Triton X-100 Surfact-Amps™ detergent solution 50 ml PIER85111
Triton X-100 Surfact-Amps™ detergent solution 250 ml PIER85112
Triton™ X-100 Surfact-Amps™ detergent solution 1.000 ml PIER28313
Triton™ X-114 Surfact-Amps™ detergent solution, 6×10 ml 1 SET PIER28332
Tween™ 20 Surfact-Amps™ detergent solution 50 ml PIER85113
Tween™ 20 Surfact-Amps™ detergent solution 250 ml PIER85114
Tween™ 20 Surfact-Amps™ detergent solution 500 ml PIER85115
Tween™ 20 Surfact-Amps™ detergent solution 1 l PIER28321
Tween™ 20 Surfact-Amps™ detergent solution, 6×10 ml 1 SET PIER28320
Tween™ 80 Surfact-Amps™ detergent solution 50 ml PIER28329
Tween™ 80 Surfact-Amps™ detergent solution 500 ml PIER28230
Tween™ 80 Surfact-Amps™ detergent solution, 6×10 ml 1 SET PIER28328
Brij™-35 Surfact-Amps™ detergent solution 50 ml PIER85117
Brij™-35 Surfact-Amps™ detergent solution 500 ml PIER85118
Brij™-35 Surfact-Amps™ detergent solution, 6×10 ml 1 SET PIER28316
Brij™-58 Surfact-Amps™ detergent solution, 6×10 ml 1 SET PIER28336
NP-40 (Nonidet P-40) Surfact-Amps™ detergent solution 500 ml PIER85125
NP-40 (Nonidet P-40) Surfact-Amps™ detergent solution, 6×10 ml 1 SET PIER28324
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal