3-Pyridylacetonitrile 98%
Varning
ACRO131970010EA 535 SEK
ACRO131970010 ACRO131970250 ACRO131970050
3-Pyridylacetonitrile 98%
3-Pyridylacetonitrile
Formel: C₇H₆N₂
MW: 118,14 g/mol
Kokpunkt: 105…108 °C (1,5 mmHg)
Densitet: 1,1 g/cm³
Flampunkt: 98 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00006406
CAS nummer: 6443-85-2
UN: 3276
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5280 to 1.5300 (20°C, 589 nm)
GC ≥97.5 %
UV at 290 nm A: ≤0.5
Titration with NaOH ≥98.5 %
Heavy metals (as Pb) ≤5 ppm