Ultra-low temperature freezer, Forma™ 900 Series
Leverantör: Thermo Scientific
Forma™
HERA902EA 0 SEK
HERA902
Ultra-low temperature freezer, Forma™ 900 Series
Forma™ 900 series freezers provides uncompromised sample protection for −50 to −86 °C applications. Ideal for universities, research facilities and industrial institutions.
  • Polyurethane foam insulation: Reduces power consumption and maintains temperature setpoint
  • Rugged, heavy gauge, cold-rolled steel cabinets resist chipping and rust
  • Centralized information center, including microprocessor control and monitoring system
  • All controls and displays are easy to reach and read
  • Ergonomic door handle with integrated key lock and padlock standard