VWR® Fluorescent Protein Quantitation Kit
Leverantör: Avantor
1B1622-KITEA 2525 SEK
VWRC1B1622-KIT
VWR® Fluorescent Protein Quantitation Kit
Proteinextraktions Protein Extraction Kits
Fluorescent Protein Quantitation Kit uses a dye to bind to primary amines of proteins and peptides in solution. It quantifies protein in the low microgram per ml (1 μg/ml) range up to 500 microgram per ml (500 μg/ml).
Leveransinformation: Kit includes 5 fluorescent protein assay buffer tablets, 1 ml of fluorescent protein dye, 2×1,0 ml of fluorescent protein standard and sufficient material for 100×2 ml reactions.