3D gloss meter
Leverantör: Hach
664-0223EA 0 SEK
664-0223
3D gloss meter
Coatingmätare
This multiangular reflectometer is designed with a three-angle geometry perfectly suited to each and every field of gloss measurement.
Suitable for metallic and non-metallic surfaces.
Certifikat: ISO 7668