Anti-IgG1 Human Antibody
Leverantör: MP Biomedicals
ICNA08642311EA 3900 SEK
ICNA08642311 ICNA08642312
Anti-IgG1 Human Antibody
Antikroppar
This antibody is purified from human myeloma sera containing high levels of IgG1 Kappa protein. The protein was purified using Ion-exchange, gel filtration and affinity chromatography.
This product contains 1% IgG2, IgG3, and IgG4 .
Type: Secondary
Antigen: IgG1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Human
Isotype:
Reactivity:
Order Now