Emergency shower outlet, for wall mounting
Leverantör: Broen Lab
118-0401EA 6700 SEK
118-0401
Emergency shower outlet, for wall mounting
Första hjälpen produkter Duschar
This body shower outlet is intended for wall mounting. Made of brass with chemical resistant BROEN polycoating.
  • Without handle or pedal
  • With self-draining shower head
  • Without valve
Depth: 670 mm
Connection: G1/2"
Min. working pressure: 1 bar
Certifikat: EN 15154-1; ANSI Z358.1. TÜV certified.