Cover glasses for slipping equipment, Menzel Gläser
Leverantör: Epredia
MENZBB024050M113EA 815 SEK
MENZBB024050M113
Cover glasses for slipping equipment, Menzel Gläser
The cover slips are suitable for in vitro diagnostic (IVD) applications.
  • Uniform quality of thickness: Pure white glass of hydrolytic class 1
  • No bubbles or flaws
  • Pre-washed