Full face mask, Promask2
Leverantör: Scott Safety
SCTS5512890EA 2175 SEK
SCTS5512890
Full face mask, Promask2
Promask2 is a twin filter full face mask providing exceptional balance and presenting an extremely low profile for close-up or restricted access working. It incorporates a wide panoramic visor to maximise the field of vision and a front mounted speech diaphragm for easy communication with colleagues.
  • Moulded in grey silicone
  • Ultra-comfortable T-bar faceseal produces an efficient seal with minimal pressure against the face
  • Airflow management system utilises the inner mask to prevent the visor from misting
  • Clear inner mask increases wearer acceptability and presents a user friendly appearance to others
This mask accepts the Pro2 range of cartridge filters, which have individual covers to maximise working life by protecting the filter media from splashes, sparks and contaminants.
Certifikat: EN 136