Goggles, X90
Leverantör: Bollé Safety
111-0159EA 265 SEK
111-0159
Goggles, X90
Korgglasögon
These small ultra-compact googles provide comfort, protection and attractive design.
  • Indirect ventilation
  • Adjustable band
  • Ball pivot strap attachment
Weight: 62 g
Certifikat: EN 166