Chemical storage cabinets, underbench, Captair® 822 B
Leverantör: Erlab
Captair®
135-2731EA 37300 SEK
135-2731 135-2733 ERLAB03IA02B0/HP ERLAB03IA02B0/K 135-2735 135-2734 ERLAB03IA02B0/BE ERLAB03IA02B0/AS ERLAB03IA02B0/F
Chemical storage cabinets, underbench, Captair® 822 B
Inredning Förvaringsskåp
822 B storage cabinet are underbench ductless storage cabinet.
  • Molecular or particulate filtration
  • 50 L storage capacity
  • Transparent door
Types of filters:
AS: For organic vapours
BE: Acid vapours
F: For formaldehyde vapours
K: For ammonia vapours
HP: HEPA H14 filter for powders storage
Energy consumption: 35 W
Drawer WxDxH: 650×580×755 mm